ENA447958838 Enamine

4-(quinolin-8-yl)benzoic acid

  • MDL number MFCD11933247
  • Cas Number : 216059-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210888
Availability details for
Quantity
  • Availability
  • Add to Cart