ENA447958839 Enamine

2-cyclopropaneamidopyridine-4-carboxylic acid

  • MDL number MFCD20320352
  • Cas Number : 1339368-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210889
Availability details for
Quantity
  • Availability
  • Add to Cart