ENA447958841 Enamine

N-(2-aminoethyl)-2,3-dichlorobenzene-1-sulfonamide

  • MDL number MFCD12085732
  • Cas Number : 1154979-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208878
Availability details for
Quantity
  • Availability
  • Add to Cart