ENA447958851 Enamine

2-amino-3-(dimethylamino)-2-methylpropanenitrile

  • MDL number MFCD16134225
  • Cas Number : 1248160-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210959
Availability details for
Quantity
  • Availability
  • Add to Cart