ENA447958852 Enamine

2-[(2-hydroxyethyl)amino]propanenitrile

  • MDL number MFCD10014401
  • Cas Number : 1018539-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210960
Availability details for
Quantity
  • Availability
  • Add to Cart