ENA447958861 Enamine

1-(4-methoxyphenyl)prop-2-en-1-ol

  • MDL number MFCD01732658
  • Cas Number : 51410-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208408
Availability details for
Quantity
  • Availability
  • Add to Cart