ENA447958862 Enamine

2-[(piperazin-1-yl)methyl]-3,4-dihydroquinazolin-4-one hydrochloride

  • MDL number MFCD04222087
  • Cas Number : 1158278-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210981
Availability details for
Quantity
  • Availability
  • Add to Cart