ENA447958867 Enamine

2-[(3-cyanophenyl)methanesulfonyl]benzoic acid

  • MDL number MFCD13362234
  • Cas Number : 1036549-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211001
Availability details for
Quantity
  • Availability
  • Add to Cart