ENA447958869 Enamine

imidazo[1,2-a]pyridin-2-yl trifluoromethanesulfonate

  • MDL number MFCD25954306
  • Cas Number : 1021019-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211012
Availability details for
Quantity
  • Availability
  • Add to Cart