ENA447958874 Enamine

5-benzyl-1H-1,2,4-triazole-3-carboxamide

  • MDL number MFCD24624870
  • Cas Number : 63970-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211040
Availability details for
Quantity
  • Availability
  • Add to Cart