ENA447958877 Enamine

1,8-naphthyridine-3-carbonitrile

  • MDL number MFCD18802345
  • Cas Number : 1142927-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211048
Availability details for
Quantity
  • Availability
  • Add to Cart