ENA447958878 Enamine

2-(4-chlorophenyl)-3-methylbutan-1-amine

  • MDL number MFCD11590304
  • Cas Number : 95349-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211049
Availability details for
Quantity
  • Availability
  • Add to Cart