ENA447958879 Enamine

1,5-dimethylpyrrolidine-2,4-dione

  • MDL number MFCD19690648
  • Cas Number : 849770-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211050
Availability details for
Quantity
  • Availability
  • Add to Cart