ENA447958880 Enamine

1H-1,2,3-benzotriazole-5-sulfonamide

  • MDL number MFCD18807984
  • Cas Number : 491612-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211051
Availability details for
Quantity
  • Availability
  • Add to Cart