ENA447958886 Enamine

2-ethyl-2,5-dihydro-1H-pyrrole

  • MDL number MFCD11053978
  • Cas Number : 1177359-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211059
Availability details for
Quantity
  • Availability
  • Add to Cart