ENA447958888 Enamine

2-cyclopropyl-5-methyl-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD16698634
  • Cas Number : 1249240-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211064
Availability details for
Quantity
  • Availability
  • Add to Cart