ENA447958890 Enamine

2-aminoquinoline-4-carbonitrile

  • MDL number MFCD13181832
  • Cas Number : 1232431-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211070
Availability details for
Quantity
  • Availability
  • Add to Cart