ENA447958891 Enamine

1-(3,4-dimethoxy-2-methylphenyl)ethan-1-one

  • MDL number MFCD19301905
  • Cas Number : 5417-20-9
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-211072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211072
Availability details for
Quantity
  • Availability
  • Add to Cart