ENA447958895 Enamine

3-[(4-bromothiophen-2-yl)methyl]-1,3-diazaspiro[4.4]nonane-2,4-dione

  • MDL number MFCD16222224
  • Cas Number : 1292607-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211083
Availability details for
Quantity
  • Availability
  • Add to Cart