ENA447958896 Enamine

3-(4-ethylphenyl)piperidine

  • MDL number MFCD17487065
  • Cas Number : 1260849-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211084
Availability details for
Quantity
  • Availability
  • Add to Cart