ENA447958900 Enamine

1-acetamido-4-phenylcyclohexane-1-carboxylic acid

  • MDL number MFCD19514834
  • Cas Number : 1497094-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211090

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211090
Availability details for
Quantity
  • Availability
  • Add to Cart