ENA447958902 Enamine

3-(bromomethyl)-1lambda6-thiane-1,1-dione

  • MDL number MFCD16556115
  • Cas Number : 1227465-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208291
Availability details for
Quantity
  • Availability
  • Add to Cart