ENA447958911 Enamine

4-chloro-6-nitrothieno[2,3-d]pyrimidine

  • MDL number MFCD20265441
  • Cas Number : 56844-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211125
Availability details for
Quantity
  • Availability
  • Add to Cart