ENA447958916 Enamine

4-phenyl-1lambda6,2-thiazolidine-1,1-dione

  • MDL number MFCD20483769
  • Cas Number : 320393-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211206
Availability details for
Quantity
  • Availability
  • Add to Cart