ENA447958917 Enamine

2-(2-amino-5-chlorobenzamido)benzoic acid

  • MDL number MFCD28397766
  • Cas Number : 59187-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211207
Availability details for
Quantity
  • Availability
  • Add to Cart