ENA447958919 Enamine

methyl 4-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD00159649
  • Cas Number : 68809-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211211
Availability details for
Quantity
  • Availability
  • Add to Cart