ENA447958927 Enamine

[(furan-2-yl)methyl](methyl)[3-(methylamino)propyl]amine

  • MDL number MFCD14655421
  • Cas Number : 1248515-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211232
Availability details for
Quantity
  • Availability
  • Add to Cart