ENA447958930 Enamine

3-fluoro-5-(hydroxymethyl)phenol

  • MDL number MFCD16249597
  • Cas Number : 1260762-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211247
Availability details for
Quantity
  • Availability
  • Add to Cart