ENA447958931 Enamine

3-chloro-1-(difluoromethyl)-1,2-dihydropyrazin-2-one

  • MDL number MFCD28397768
  • Cas Number : 1603237-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211256
Availability details for
Quantity
  • Availability
  • Add to Cart