ENA447958932 Enamine

5-bromo-4-methyl-2,3-dihydro-1,3-benzoxazol-2-one

  • MDL number MFCD22560731
  • Cas Number : 1388042-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211266
Availability details for
Quantity
  • Availability
  • Add to Cart