ENA447958936 Enamine

3-(phenylcarbamoyl)benzoic acid

  • MDL number MFCD00625702
  • Cas Number : 16776-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211277
Availability details for
Quantity
  • Availability
  • Add to Cart