ENA447958940 Enamine

3-phenyl-octahydrocyclopenta[b]pyrrole

  • MDL number MFCD09035108
  • Cas Number : 1053170-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208414
Availability details for
Quantity
  • Availability
  • Add to Cart