ENA447958953 Enamine

3-bromopropanamide

  • MDL number MFCD00143100
  • Cas Number : 6320-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-211425
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart