ENA447958992 Enamine

1-bromo-2-methanesulfonylethane

  • MDL number MFCD08274926
  • Cas Number : 16523-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-211467
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart