ENA447959042 Enamine

2-(5-phenyl-1,3,4-oxadiazol-2-yl)ethan-1-ol

  • MDL number MFCD28397776
  • Cas Number : 1537268-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213073
Availability details for
Quantity
  • Availability
  • Add to Cart