ENA447959046 Enamine

3-(bromomethyl)-1,2-oxazole

  • MDL number MFCD08669520
  • Cas Number : 76632-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22115
Availability details for
Quantity
  • Availability
  • Add to Cart