ENA447959066 Enamine

2-(2,6-dimethylpiperidin-1-yl)-N'-hydroxyethanimidamide

  • Cas Number : 1016846-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43420
Availability details for
Quantity
  • Availability
  • Add to Cart