ENA447959092 Enamine

5-methylthiophene-3-carboxylic acid

  • MDL number MFCD01680229
  • Cas Number : 19156-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-132
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62620
Availability details for
Quantity
  • Availability
  • Add to Cart