ENA447959095 Enamine

1-methyl-1H-imidazo[4,5-c]pyridine

  • MDL number MFCD13175881
  • Cas Number : 5028-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63857
Availability details for
Quantity
  • Availability
  • Add to Cart