ENA447959120 Enamine

methyl 5-(chloromethyl)-1,2-oxazole-4-carboxylate

  • MDL number MFCD20547602
  • Cas Number : 1027781-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74364
Availability details for
Quantity
  • Availability
  • Add to Cart