ENA447959121 Enamine

3-(4-methoxyphenyl)morpholine

  • MDL number MFCD09901532
  • Cas Number : 1017481-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-55
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74371
Availability details for
Quantity
  • Availability
  • Add to Cart