ENA447959132 Enamine

benzyl 3-(chlorosulfonyl)pyrrolidine-1-carboxylate

  • MDL number MFCD11501253
  • Cas Number : 1035173-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78397
Availability details for
Quantity
  • Availability
  • Add to Cart