ENA447959149 Enamine

4-(-lambda5-diazynylidene)-4H-imidazole-5-carboxamide

  • MDL number MFCD01721724
  • Cas Number : 7008-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84855
Availability details for
Quantity
  • Availability
  • Add to Cart