ENA447959167 Enamine

3-methoxyquinoline-4-carboxylic acid

  • MDL number MFCD21649046
  • Cas Number : 1241386-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93473
Availability details for
Quantity
  • Availability
  • Add to Cart