ENA447959175 Enamine

methyl 3-oxocyclohexane-1-carboxylate

  • MDL number MFCD09031996
  • Cas Number : 13148-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96130
Availability details for
Quantity
  • Availability
  • Add to Cart