ENA447959178 Enamine

1-tert-butyl-1H-pyrrole

  • MDL number MFCD00015558
  • Cas Number : 24764-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96181
Availability details for
Quantity
  • Availability
  • Add to Cart