ENA447959181 Enamine

5-amino-2-chlorobenzene-1-thiol

  • MDL number MFCD20694766
  • Cas Number : 97094-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97284
Availability details for
Quantity
  • Availability
  • Add to Cart