ENA447959187 Enamine

ethyl 1-methyl-1H-1,2,3-triazole-5-carboxylate

  • MDL number MFCD20922005
  • Cas Number : 137156-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99471
Availability details for
Quantity
  • Availability
  • Add to Cart