ENA447959188 Enamine

1-[2-fluoro-6-(pyrrolidin-1-yl)phenyl]methanamine

  • MDL number MFCD21881942
  • Cas Number : 1368808-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99544
Availability details for
Quantity
  • Availability
  • Add to Cart