ENA448967759 Enamine

benzyl 2-[(chlorosulfonyl)methyl]morpholine-4-carboxylate

  • MDL number MFCD28505645
  • Cas Number : 1803584-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136399

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136399
Availability details for
Quantity
  • Availability
  • Add to Cart